Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(9H-Carbazol-9-yl)aniline hydrochloride 95%

Supplier: AMBEED, INC
Warning

A628635-100MG A628635-1G A628635-250MG
76696-776EA 13 USD InStock
76696-776 76696-778 76696-780
4-(9H-Carbazol-9-yl)aniline hydrochloride 95%
4-(9H-Carbazol-9-yl)aniline hydrochloride


Formula: C₁₈H₁₅ClN₂
MDL Number: MFCD00082678
CAS Number: 312700-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty