Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltriethylammonium tetrafluoroborate 98+%

Supplier: AMBEED, INC
Warning

A631140-100G A631140-25G A631140-5G
76696-788EA 174 USD InStock
76696-788 76696-790 76696-792
Benzyltriethylammonium tetrafluoroborate 98+%
Benzyltriethylammonium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₃H₂₂BF₄N
MDL Number: MFCD00011823
CAS Number: 77794-93-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty