Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)thiomorpholine-1,1-dioxide 98%

Supplier: AMBEED, INC
Warning

A646132-1G A646132-250MG A646132-5G
76696-942EA 18 USD InStock
76696-942 76696-944 76696-946
4-(4-Chlorophenyl)thiomorpholine-1,1-dioxide 98%
4-(4-Chlorophenyl)thiomorpholine-1,1-dioxide
Precaución: for Research Use Only


Formula: C₁₀H₁₂ClNO₂S
MW: 245.72 g/mol
MDL Number: MFCD07366283
CAS Number: 82222-74-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty