Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Norbornane-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A655635-100G A655635-10G A655635-1G A655635-25G A655635-5G
76697-000EA 416 USD InStock
76697-000 76697-002 76697-004 76697-006 76697-008
Norbornane-2-carboxylic acid 98%
Norbornane-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD00167749
CAS Number: 824-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty