Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (1R,2S)-1-amino-2-vinylcyclopropane-1-carboxylate hydrochloride 97+%

Supplier: AMBEED, INC
Warning

A657661-100MG A657661-250MG
76697-024EA 45 USD InStock
76697-024 76697-026
Ethyl (1R,2S)-1-amino-2-vinylcyclopropane-1-carboxylate hydrochloride 97+%
Ethyl (1R,2S)-1-amino-2-vinylcyclopropane-1-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₄ClNO₂
MDL Number: MFCD11042409
CAS Number: 259214-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty