Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Het anhydride 98%

Supplier: AMBEED, INC
Warning

A674320-100G A674320-500G
76697-154EA 7 USD InStock
76697-154 76697-156
Het anhydride 98%
Het anhydride
Precaución: for Research Use Only


Formula: C₉H₂Cl₆O₃
MW: 370.81 g/mol
MDL Number: MFCD00080438
CAS Number: 115-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty