Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzyl adipate 95%

Supplier: AMBEED, INC
Warning

A701138-1G A701138-5G
76697-318EA 13 USD InStock
76697-318 77602-784
Dibenzyl adipate 95%
Dibenzyl adipate


Formula: C₂₀H₂₂O₄
MDL Number: MFCD00059667
CAS Number: 2451-84-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty