Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Morpholino benzoate 95%

Supplier: AMBEED, INC
Warning

A704856-100MG A704856-1G A704856-250MG A704856-5G
76697-342EA 26 USD InStock
76697-342 76697-344 76697-346 76697-348
Morpholino benzoate 95%
Morpholino benzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₃
MW: 207.22 g/mol
MDL Number: MFCD09032166
CAS Number: 5765-65-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty