Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acid green 1 98%

Supplier: AMBEED, INC
Warning

A709376-1000G A709376-100G A709376-25G A709376-500G
76697-376EA 109 USD InStock
76697-376 76697-378 76697-380 76697-382
Acid green 1 98%
Acid green 1
Precaución: for Research Use Only


Formula: C₃₀H₁₈FeN₃O₁₅S₃
MW: 878.45 g/mol
MDL Number: MFCD00003886
CAS Number: 19381-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty