Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-2-(4-ethoxyphenyl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A710593-100MG A710593-1G A710593-250MG A710593-5G
76697-394EA 11 USD InStock
76697-394 76697-396 76697-398 76697-400
2-Amino-2-(4-ethoxyphenyl)acetic acid 95%
2-Amino-2-(4-ethoxyphenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00665360
CAS Number: 299168-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty