Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-5-methylsalicylate 98%

Supplier: AMBEED, INC
Warning

A716356-100G A716356-10G A716356-1G A716356-25G A716356-5G
76697-438EA 380 USD InStock
76697-438 76697-440 76697-442 76697-444 76697-446
Methyl-5-methylsalicylate 98%
Methyl 5-methylsalicylate
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MDL Number: MFCD00016485
CAS Number: 22717-57-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty