Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pararosaniline 98%

Supplier: AMBEED, INC
Warning

A730641-1G A730641-25G A730641-5G
76697-592EA 12 USD InStock
76697-592 76697-594 76697-596
Pararosaniline 98%
Pararosaniline
Precaución: for Research Use Only


Formula: C₁₉H₁₉N₃O
MDL Number: MFCD00036222
CAS Number: 467-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty