Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dimethoxyphenylacetylene 98%

Supplier: AMBEED, INC
Warning

A734359-1G A734359-250MG A734359-25G A734359-5G
76697-614EA 19 USD InStock
76697-614 76697-616 76697-618 76697-620
3,4-Dimethoxyphenylacetylene 98%
3,4-Dimethoxyphenylacetylene
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MW: 162.185 g/mol
MDL Number: MFCD03839986
CAS Number: 4302-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty