Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,3R,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate 98%

Supplier: AMBEED, INC
Warning

A736080-100G A736080-1G A736080-250MG A736080-25G A736080-5G
76697-622EA 207 USD InStock
76697-622 76697-624 76697-626 76697-628 76697-630
(2R,3R,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate 98%
(2R,3R,4S,5R,6R)-6-((Benzoyloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetrabenzoate
Precaución: for Research Use Only


Formula: C₄₁H₃₂O₁₁
MW: 700.68 g/mol
MDL Number: MFCD00010695
CAS Number: 22415-91-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty