Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(tetramethylene)fluoroformamidinium hexafluorophosphate 99%

Supplier: AMBEED, INC
Danger

A741194-10G A741194-1G A741194-250MG A741194-25G A741194-5G
76697-662EA 23 USD InStock
76697-662 76697-664 76697-666 76697-668 76697-670
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate 99%
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₉H₁₆F₇N₂P
MDL Number: MFCD02683430
CAS Number: 164298-25-3
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty