Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltrimethylsilane 96%

Supplier: AMBEED, INC
Danger

A743996-1G A743996-250MG A743996-5G
76697-680EA 6 USD InStock
76697-680 76697-682 76697-684
Benzyltrimethylsilane 96%
Benzyltrimethylsilane
Precaución: for Research Use Only


Formula: C₁₀H₁₆Si
MDL Number: MFCD00039796
CAS Number: 770-09-2
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty