Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-L-alanine 4-nitrophenyl ester 98%

Supplier: AMBEED, INC
Warning

A744170-1G A744170-25G A744170-5G
76697-686EA 10 USD InStock
76697-686 76697-688 76697-690
N-Boc-L-alanine 4-nitrophenyl ester 98%
N-Boc-L-alanine 4-nitrophenyl ester
Precaución: for Research Use Only


Formula: C₁₄H₁₈N₂O₆
MW: 310.306 g/mol
MDL Number: MFCD00038117
CAS Number: 2483-49-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty