Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Isopropyl-2-methoxybenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A746332-1G A746332-250MG A746332-5G
76697-696EA 20 USD InStock
76697-696 76697-698 76697-700
5-Isopropyl-2-methoxybenzeneboronic acid 98%
5-Isopropyl-2-methoxybenzeneboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₃
MDL Number: MFCD01318154
CAS Number: 216393-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty