Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(II) trifluoroacetate hydrate 95%

Supplier: AMBEED, INC
Warning

A749193-100G A749193-25G A749193-5G
76697-714EA 95 USD InStock
76697-714 76697-716 76697-718
Copper(II) trifluoroacetate hydrate 95%
Copper(II) trifluoroacetate hydrate
Precaución: for Research Use Only


Formula: C₄CuF₆O₄·xH₂O
MW: 289.576 g/mol (anhy)
MDL Number: MFCD00013200
CAS Number: 123333-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty