Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzyl isothiocyanate 97%

Supplier: AMBEED, INC
Danger

A761319-25G A761319-5G
76697-762EA 68 USD InStock
76697-762 76697-764
4-Fluorobenzyl isothiocyanate 97%
4-Fluorobenzyl isothiocyanate
Precaución: for Research Use Only


Formula: C₈H₆FNS
MW: 167.21 g/mol
Boiling Pt: 90
MDL Number: MFCD00041115
CAS Number: 2740-88-7
UN: 2922
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty