Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Butyl-4-methylpyridinium bromide 97%

Supplier: AMBEED, INC
Warning

A769056-100G A769056-1G A769056-25G A769056-5G
76697-810EA 117 USD InStock
76697-810 76697-812 76697-814 76697-816
1-Butyl-4-methylpyridinium bromide 97%
1-Butyl-4-methylpyridinium bromide
Precaución: for Research Use Only


Formula: C₁₀H₁₆BrN
MW: 230.15 g/mol
MDL Number: MFCD06798209
CAS Number: 65350-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty