Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cetyldimethylethylammonium bromide 99%

Supplier: AMBEED, INC
Warning

A774734-100G A774734-25G
76697-844EA 48 USD InStock
76697-844 76697-846
Cetyldimethylethylammonium bromide 99%
Cetyldimethylethylammonium bromide
Precaución: for Research Use Only


Formula: C₂₀H₄₄BrN
MDL Number: MFCD00011774
CAS Number: 124-03-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty