Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl (±)-lactate 98%

Supplier: AMBEED, INC
Warning

A776059-100G A776059-25G A776059-500G
76697-854EA 23 USD InStock
76697-854 76697-856 76697-858
Butyl (±)-lactate 98%
Butyl (±)-lactate
Precaución: for Research Use Only


Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 170
Melting Pt: -28
MDL Number: MFCD00004519
CAS Number: 138-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty