Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium adipate 97%

Supplier: AMBEED, INC
Warning

A781154-100G A781154-500G A781154-1000G
76697-870EA 7 USD InStock
76697-870 76697-872 77627-546
Ammonium adipate 97%
Ammonium adipate
Precaución: for Research Use Only


Formula: C₆H₁₆N₂O₄
MW: 180.20 g/mol
MDL Number: MFCD00210236
CAS Number: 19090-60-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty