Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-methylbenzotrifluoride 98%

Supplier: AMBEED, INC
Warning

A786022-1G A786022-25G A786022-5G A786022-100G
76697-940EA 9 USD InStock
76697-940 76697-942 76697-944 77627-572
5-Amino-2-methylbenzotrifluoride 98%
5-Amino-2-methylbenzotrifluoride
Precaución: for Research Use Only


Formula: C₈H₈F₃N
MDL Number: MFCD01631582
CAS Number: 65934-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty