Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(4-bromophenyl)disulfane 98%

Supplier: AMBEED, INC
Danger

A787378-250MG A787378-5G A787378-1G
76697-948EA 16 USD InStock
76697-948 76697-950 77627-576
1,2-Bis(4-bromophenyl)disulfane 98%
1,2-Bis(4-bromophenyl)disulfane
Precaución: for Research Use Only


Formula: C₁₂H₈Br₂S₂
MW: 376.12 g/mol
MDL Number: MFCD00017832
CAS Number: 5335-84-2
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty