Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Nitrophenyl)butanoic acid 98%

Supplier: AMBEED, INC
Warning

A795621-100G A795621-1G A795621-25G A795621-5G
76697-988EA 157 USD InStock
76697-988 76697-990 76697-992 76697-994
2-(4-Nitrophenyl)butanoic acid 98%
2-(4-Nitrophenyl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00156935
CAS Number: 7463-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty