Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ac-Trp-3,5-bis(trifluoromethyl)benzyl ester 98%

Supplier: AMBEED, INC
Warning

A801868-1MG A801868-5MG
76698-050EA 11 USD InStock
76698-050 76698-052
Ac-Trp-3,5-bis(trifluoromethyl)benzyl ester 98%
Ac-Trp-3,5-bis(trifluoromethyl)benzyl ester
Precaución: for Research Use Only


Formula: C₂₂H₁₈F₆N₂O₃
MW: 472.39 g/mol
MDL Number: MFCD00237267
CAS Number: 148451-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty