Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methylanisole 95%

Supplier: AMBEED, INC
Warning

A825350-100G A825350-1G A825350-25G A825350-5G
76698-202EA 101 USD InStock
76698-202 76698-204 76698-206 76698-208
4-Fluoro-3-methylanisole 95%
4-Fluoro-3-methylanisole
Precaución: for Research Use Only


Formula: C₈H₉FO
MDL Number: MFCD00042293
CAS Number: 2338-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty