Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-3-nitro-4-quinolinol 97%

Supplier: AMBEED, INC
Warning

A830711-1G A830711-250MG A830711-25G A830711-5G
76698-250EA 32 USD InStock
76698-250 76698-252 76698-254 76698-256
7-Chloro-3-nitro-4-quinolinol 97%
7-Chloro-3-nitro-4-quinolinol
Precaución: for Research Use Only


Formula: C₉H₅ClN₂O₃
MW: 224.6006 g/mol
MDL Number: MFCD09800417
CAS Number: 5350-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty