Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Phenylethyl propionate 98%

Supplier: AMBEED, INC
Warning

A841618-1000G A841618-100G A841618-25G A841618-500G
76698-328EA 147 USD InStock
76698-328 76698-330 76698-332 76698-334
1-Phenylethyl propionate 98%
1-Phenylethyl propionate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MDL Number: MFCD00009305
CAS Number: 120-45-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty