Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3'-Trifluoromethoxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A844383-10G A844383-1G A844383-25G A844383-500G A844383-100G
76698-342EA 19 USD InStock
76698-342 76698-344 76698-346 76698-348 77627-648
3'-Trifluoromethoxyacetophenone 98%
3'-Trifluoromethoxyacetophenone
Precaución: for Research Use Only


Formula: C₉H₇F₃O₂
MW: 204.15
MDL Number: MFCD00042403
CAS Number: 170141-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty