Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexyl sulphide 90%

Supplier: AMBEED, INC
Warning

A880641-100G A880641-25G
76698-508EA 59 USD InStock
76698-508 76698-510
Dicyclohexyl sulphide 90%
Dicyclohexyl sulphide
Precaución: for Research Use Only


Formula: C₁₂H₂₂S₂
MW: 230.44 g/mol
MDL Number: MFCD00013759
CAS Number: 2550-40-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty