Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cobalt(II) Phthalocyanine 96%

Supplier: AMBEED, INC
Danger

A895711-1G A895711-25G A895711-5G
76698-566EA 17 USD InStock
76698-566 76698-568 76698-570
Cobalt(II) Phthalocyanine 96%
Cobalt(II) Phthalocyanine
Precaución: for Research Use Only


Formula: C₃₂H₁₆CoN₈
MW: 571.47 g/mol
MDL Number: MFCD00010718
CAS Number: 3317-67-7
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty