Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium 3-hydroxy-3-methylbutanoate 96%

Supplier: AMBEED, INC
Warning

A902202-100G A902202-5G
76698-614EA 28 USD InStock
76698-614 76698-616
Calcium 3-hydroxy-3-methylbutanoate 96%
Calcium 3-hydroxy-3-methylbutanoate


Formula: C₁₀H₁₈CaO₆
MW: 274.32 g/mol
MDL Number: MFCD01318562
CAS Number: 135236-72-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty