Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyloxy-tert-butyldimethylsilane 97%

Supplier: AMBEED, INC
Danger

A922795-1G A922795-25G
76698-710EA 13 USD InStock
76698-710 76698-712
Allyloxy-tert-butyldimethylsilane 97%
Allyloxy-tert-butyldimethylsilane
Precaución: for Research Use Only


Formula: C₉H₂₀OSi
MDL Number: MFCD00239355
CAS Number: 105875-75-0
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty