Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cinnamyl cinnamate 98%

Supplier: AMBEED, INC
Warning

A923084-100G A923084-25G A923084-5G
76698-714EA 58 USD InStock
76698-714 76698-716 76698-718
Cinnamyl cinnamate 98%
Cinnamyl cinnamate
Precaución: for Research Use Only


Formula: C₁₈H₁₆O₂
MW: 264.32 g/mol
MDL Number: MFCD00037826
CAS Number: 122-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty