Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Nitrophenyl)butanoic acid 99+%

Supplier: AMBEED, INC
Warning

A946881-100G A946881-1G A946881-5G
76698-802EA 435 USD InStock
76698-802 76698-804 76698-806
4-(4-Nitrophenyl)butanoic acid 99+%
4-(4-Nitrophenyl)butanoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00007387
CAS Number: 5600-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty