Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibutyryl-cAMP sodium salt 98%

Supplier: AMBEED, INC
Warning

A976872-100MG A976872-10MG A976872-1MG A976872-250MG A976872-50MG A976872-1G
76698-888EA 53 USD InStock
76698-888 76698-890 76698-892 76698-894 76698-896 77603-376
Dibutyryl-cAMP sodium salt 98%
Dibutyryl-cAMP sodium salt
Precaución: for Research Use Only


Formula: C₁₈H₂₃N₅O₈P·Na
MW: 491.37 g/mol
MDL Number: MFCD00005843
CAS Number: 16980-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty