Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Hydroxybiphenyl-4-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A102746-100G A102746-10G A102746-1G A102746-25G A102746-5G
76717-652EA 135 USD InStock
76717-652 76717-654 76717-656 76717-658 77666-480
4'-Hydroxybiphenyl-4-carboxylic acid 98%
4'-Hydroxybiphenyl-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
MDL Number: MFCD00059078
CAS Number: 58574-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty