Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-D-pantothenate 95%

Supplier: AMBEED, INC
Warning

A105231-100MG A105231-25G
76717-754EA 5 USD InStock
76717-754 76717-756 76717-758 76717-760
Sodium-D-pantothenate 95%
Sodium-D-pantothenate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₅
MW: 241.22 g/mol
MDL Number: MFCD00002767
CAS Number: 867-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty