Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-amino-1-methyl-1H-pyrazole-5-carboxylate 95%

Supplier: AMBEED, INC
Warning

A105308-100MG A105308-250MG A105308-1G
76717-762EA 18 USD InStock
76717-762 76717-764 77666-542
Methyl 3-amino-1-methyl-1H-pyrazole-5-carboxylate 95%
Methyl 3-amino-1-methyl-1H-pyrazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂
MDL Number: MFCD08443798
CAS Number: 89088-56-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty