Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Dimethylamino)-2-oxoacetic acid 97%

Supplier: AMBEED, INC
Danger

A105385-100G A105385-1G A105385-25G A105385-500G A105385-5G
76717-770EA 72 USD InStock
76717-770 76717-772 76717-774 76717-776 76717-778
2-(Dimethylamino)-2-oxoacetic acid 97%
2-(Dimethylamino)-2-oxoacetic acid
Precaución: for Research Use Only


Formula: C₄H₇NO₃
MDL Number: MFCD01971023
CAS Number: 32833-96-8
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty