Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-2-bromobenzylamine 98%

Supplier: AMBEED, INC
Warning

A106689-1G A106689-5G
76717-870EA 34 USD InStock
76717-870 76717-872
N-Boc-2-bromobenzylamine 98%
N-Boc-2-bromobenzylamine
Precaución: for Research Use Only


Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD05664385
CAS Number: 162356-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty