Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium acrylate 98% (by titrimetric analysis) (~white)

Supplier: AMBEED, INC
Danger

A107105-10G A107105-25G A107105-5G
76717-880EA 14 USD InStock
76717-880 76717-882 76717-884
Sodium acrylate 98% (by titrimetric analysis) (~white)
Sodium acrylate
Precaución: for Research Use Only


Formula: C₃H₃NaO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00067207
CAS Number: 7446-81-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty