Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rolipram 98%

Supplier: AMBEED, INC
Warning

A110225-10MG A110225-1MG A110225-50MG A110225-1G A110225-250MG A110225-5G
76718-048EA 17 USD InStock
76718-048 76718-050 76718-052 77666-608 77666-610 77666-612
Rolipram 98%
Rolipram
Precaución: for Research Use Only


Formula: C₁₆H₂₁NO₃
MDL Number: MFCD00270906
CAS Number: 61413-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty