Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyl carbanilide 95%

Supplier: AMBEED, INC
Warning

A114940-100G A114940-10G A114940-1G A114940-250MG A114940-25G A114940-5G
76718-258EA 172 USD InStock
76718-258 76718-260 76718-262 76718-264 76718-266 76718-268
N,N-Dimethyl carbanilide 95%
N,N-Dimethyl carbanilide
Precaución: for Research Use Only


Formula: C₁₅H₁₆N₂O
MW: 240.306 g/mol
MDL Number: MFCD00025642
CAS Number: 611-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty