Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium glycinate 98%

Supplier: AMBEED, INC
Warning

A116718-100G A116718-25G A116718-5G A116718-500G
76718-340EA 21 USD InStock
76718-340 76718-342 76718-344 77599-686
Sodium glycinate 98%
Sodium glycinate
Precaución: for Research Use Only


Formula: C₂H₄NNaO₂
MW: 97.049 g/mol
MDL Number: MFCD00062543
CAS Number: 6000-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty