Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-hydroxy-3-iodobenzoate 97%

Supplier: AMBEED, INC
Danger

A124457-100G A124457-10G A124457-1G A124457-250MG A124457-25G A124457-5G
76718-670EA 144 USD InStock
76718-670 76718-672 76718-674 76718-676 76718-678 76718-680
Methyl-4-hydroxy-3-iodobenzoate 97%
Methyl-4-hydroxy-3-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₇IO₃
MW: 278.04 g/mol
MDL Number: MFCD05664378
CAS Number: 15126-06-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty