Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrophenyl butyrate 97%

Supplier: AMBEED, INC
Warning

A125811-10G A125811-1G A125811-5G
76718-718EA 187 USD InStock
76718-718 76718-720 76718-722
2-Nitrophenyl butyrate 97%
2-Nitrophenyl butyrate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00036217
CAS Number: 2487-26-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty